missing translation for 'onlineSavingsMsg'
Learn More

EFTUD1 Antibody, Novus Biologicals™

Product Code. 18397954 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397954 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397954 Supplier Novus Biologicals Supplier No. NBP318400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EFTUD1 Polyclonal antibody specifically detects EFTUD1 in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFTUD1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Immunoprecipitation 15-20 ug/mg lysate
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias EFL1, Elongation Factor Tu GTP Binding Domain Containing 1, Elongation Factor Tu GTP-Binding Domain-Containing Protein 1, Elongation Factor-Like 1, FAM42A, HsT19294, Protein FAM42A, RIA1, Ribosome Assembly 1 Homolog, Ribosome Assembly 1 Homolog (Yeast)
Host Species Rabbit
Immunogen Recognizes region between residues Between between 50 and 100..
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79631
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.