missing translation for 'onlineSavingsMsg'
Learn More

Efonidipine hydrochloride monoethanolate, MedChemExpress

Product Code. p-200095993
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16455218

Brand: Medchem Express HY12502A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Efonidipine hydrochloride monoethanolate (NZ-105 hydrochloride monoethanolate) is a dual T-type and L-type calcium channel blocker (CCB).

Efonidipine hydrochloride monoethanolate (NZ-105 hydrochloride monoethanolate) is a dual T-type and L-type calcium channel blocker (CCB).

Specifications

Chemical Name or Material Efonidipine hydrochloride monoethanolate
CAS 111011-76-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C36H45ClN3O8P
Quantity 10 mg
Synonym NZ-105 hydrochloride monoethanolate
Solubility Information DMSO : 125 mg/mL (175.03 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC1=C(P2(OCC(C)(C)CO2)=O)C(C3=CC([N+]([O-])=O)=CC=C3)C(C(OCCN(C4=CC=CC=C4)CC5=CC=CC=C5)=O)=C(C)N1.[H]Cl.CCO
Molecular Weight (g/mol) 714.18
Formula Weight 714.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.