missing translation for 'onlineSavingsMsg'
Learn More

Efonidipine hydrochloride monoethanolate, MedChemExpress

Product Code. 16455218
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455218 10 mg 10mg
16411437 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16455218 Supplier Medchem Express Supplier No. HY12502A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Efonidipine hydrochloride monoethanolate (NZ-105 hydrochloride monoethanolate) is a dual T-type and L-type calcium channel blocker (CCB).

Efonidipine hydrochloride monoethanolate (NZ-105 hydrochloride monoethanolate) is a dual T-type and L-type calcium channel blocker (CCB).

Specifications

Chemical Name or Material Efonidipine hydrochloride monoethanolate
CAS 111011-76-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C36H45ClN3O8P
Quantity 10 mg
Synonym NZ-105 hydrochloride monoethanolate
Solubility Information DMSO : 125 mg/mL (175.03 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC1=C(P2(OCC(C)(C)CO2)=O)C(C3=CC([N+]([O-])=O)=CC=C3)C(C(OCCN(C4=CC=CC=C4)CC5=CC=CC=C5)=O)=C(C)N1.[H]Cl.CCO
Molecular Weight (g/mol) 714.18
Formula Weight 714.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.