missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eflornithine Hydrochloride Monohydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Difluoromethylornithine hydrochloride monohydrate |
| CAS | 96020-91-6 |
| Molecular Formula | C6 H12 F2 N2 O2 . Cl H . H2 O |
| Quantity | 1 g |
| Synonym | Ornithine, 2-(difluoromethyl)-, hydrochloride, hydrate (1:1:1), DL-Ornithine, 2-(difluoromethyl)-, monohydrochloride, monohydrate, Ornithine, 2-(difluoromethyl)-, monohydrochloride, monohydrate (9CI), Eflornithine hydrochloride monohydrate |
| InChI Formula | InChI=1S/C6H12F2N2O2.ClH.H2O/c7-4(8)6(10,5(11)12)2-1-3-9;;/h4H,1-3,9-10H2,(H,11,12);1H;1H2 |
| SMILES | O.Cl.NCCCC(N)(C(F)F)C(=O)O |
| IUPAC Name | 2,5-diamino-2-(difluoromethyl)pentanoic acid;hydrate;hydrochloride |
| Molecular Weight (g/mol) | 236.64 |
| Formula Weight | 236.0739 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?