missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eflornithine hydrochloride monohydrate

Product Code. 15801052 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15801052

Brand: Thermo Scientific Chemicals 462700010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96020-91-6 | C6H15ClF2N2O3 | 236.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96020-91-6
Molecular Formula C6H15ClF2N2O3
Molecular Weight (g/mol) 236.64
MDL Number MFCD00242734
InChI Key FJPAMFNRCFEGSD-UHFFFAOYNA-N
IUPAC Name hydrogen 2,5-diamino-2-(difluoromethyl)pentanoic acid hydrate chloride
SMILES [H+].O.[Cl-].NCCCC(N)(C(F)F)C(O)=O

Specifications

Quantity 1 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: slightly soluble in ethanol
Formula Weight 236.64
Chemical Name or Material Eflornithine hydrochloride monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.