missing translation for 'onlineSavingsMsg'
Learn More

Eflornithine hydrochloride hydrate, MedChemExpress

Product Code. 16498436
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16498436

Brand: Medchem Express HYB0744B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eflornithine hydrochloride hydrate (DFMO hydrochloride hydrate) is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. Eflornithine hydrochloride hydrate is a medication for the treatment of African trypanosomiasis and excessive facial hair growth in women.

Eflornithine hydrochloride hydrate (DFMO hydrochloride hydrate) is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. Eflornithine hydrochloride hydrate is a medication for the treatment of African trypanosomiasis and excessive facial hair growth in women.

Specifications

Chemical Name or Material Eflornithine hydrochloride hydrate
Physical Form Solid
CAS 96020-91-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H15ClF2N2O3
For Use With (Application) Cancer-programmed cell death
Synonym DFMO hydrochloride hydrate MDL-71782 hydrochloride hydrate RMI-71782 hydrochloride hydrate α-difluoromethylornithine hydrochloride hydrate
Solubility Information H2O : 83.33 mg/mL (352.14 mM; Need ultrasonic)
SMILES NC(CCCN)(C(F)F)C(O)=O.[H]Cl.O
Molecular Weight (g/mol) 236.64
Formula Weight 236.64
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.