missing translation for 'onlineSavingsMsg'
Learn More

Efinaconazole-N-oxide, TRC

Product Code. 30385655
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385655 10 mg
30381860 50 mg
30378245 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385655 Supplier Toronto Research Chemicals Supplier No. TRCE43509010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2055038-63-4
Molecular Formula C18H22F2N4O2
Molecular Weight (g/mol) 364.39
Synonym (αR,βR)-α-(2,4-Difluorophenyl)-β-methyl-4-methylene-α-(1H-1,2,4-triazol-1-ylmethyl)-1-piperidineethanol 1-Oxide, 1-((2R,3R)-3-(2,4-Difluorophenyl)-3-hydroxy-4-(1H-1,2,4-triazol-1-yl)butan-2-yl)-4-methylenepiperidine 1-Oxide
SMILES O[C@](CN1N=CN=C1)([C@H](N2(CCC(CC2)=C)=O)C)C3=C(F)C=C(F)C=C3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H22F2N4O2/c1-13-5-7-24(26,8-6-13)14(2)18(25,10-23-12-21-11-22-23)16-4-3-15(19)9-17(16)20/h3-4,9,11-12,14,25H,1,5-8,10H2,2H3/t14-,18-/m1/s1
Formula Weight 364.1711
Percent Purity >95%
Chemical Name or Material Efinaconazole-N-oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.