missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFHD2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92752-0.1ml
This item is not returnable.
View return policy
Description
EFHD2 Polyclonal antibody specifically detects EFHD2 in Mouse samples. It is validated for Western Blot
Specifications
| EFHD2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| EF hand domain containing 2, EF hand domain family, member D2, EF-hand domain family, member D2, EF-hand domain-containing protein D2, MGC4342, swiprosin 1, swiprosin-1, SWS1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-78 of human EFHD2 (NP_077305.2). MATDELATKLSRRLQMEGEGGGETPEQPGLNGAAAAAAGAPDEAAEALGSADCELSAKLLRRADLNQGIGEPQSPSRR | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 79180 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction