missing translation for 'onlineSavingsMsg'
Learn More

EFHD1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18419944 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419944 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419944 Supplier Novus Biologicals Supplier No. NBP177134MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EFHD1 Polyclonal antibody specifically detects EFHD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFHD1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp781H0842, EF-hand domain family, member D1, EF-hand domain-containing protein 1, EF-hand domain-containing protein D1, member D1, MST133, MSTP133, Swiprosin-2
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human EFHD1. The immunogen is located within the first 50 amino acids of EFHD1. Amino Acid Squence: EPKPEPEPPARAPT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 80303
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.