missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFHD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£141.00
Specifications
| Antigen | EFHD1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
EFHD1 Polyclonal specifically detects EFHD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| EFHD1 | |
| Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9BUP0 | |
| 80303 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human EFHD1. The immunogen is located within the first 50 amino acids of EFHD1. Amino Acid Squence: EPKPEPEPPARAPT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| DKFZp781H0842, EF-hand domain family, member D1, EF-hand domain-containing protein 1, EF-hand domain-containing protein D1, member D1, MST133, MSTP133, Swiprosin-2 | |
| EFHD1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title