missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFHD1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77134AF594
This item is not returnable.
View return policy
Description
EFHD1 Polyclonal antibody specifically detects EFHD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| EFHD1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 594 | |
| DKFZp781H0842, EF-hand domain family, member D1, EF-hand domain-containing protein 1, EF-hand domain-containing protein D1, member D1, MST133, MSTP133, Swiprosin-2 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human EFHD1. The immunogen is located within the first 50 amino acids of EFHD1. Amino Acid Squence: EPKPEPEPPARAPT | |
| 0.1 mL | |
| Neuroscience | |
| 80303 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction