missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EFHC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13426137
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13426137 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13426137 Supplier Invitrogen™ Supplier No. PA531608

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

EFHC2 (EF-hand domain-containing protein 2) is a gene located on the X chromosome at position Xp11.4. The protein encoded by EFHC2 is characterized by the presence of EF-hand motifs, which are helix-loop-helix structural domains capable of binding calcium ions. The EFHC2 protein is involved in various cellular processes, particularly those that are calcium-dependent, such as signal transduction and regulation of enzyme activities. EFHC2 has been implicated in neurological functions and conditions. For instance, it has been studied in the context of epilepsy, where variants in the gene are associated with increased susceptibility to juvenile myoclonic epilepsy. Additionally, EFHC2 is expressed in the brain and testis, suggesting roles in neurodevelopment and reproduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFHC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene EFHC2
Gene Accession No. Q5JST6
Gene Alias 4933407D04Rik; dJ1158H2.1; EF-hand domain (C-terminal) containing 2; EF-hand domain containing 2; EF-hand domain-containing family member C2; Efhc2; mental retardation, X-linked 74; mRib72-2; MRX74
Gene Symbols EFHC2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EFHC2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80258
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.