Learn More
EFHC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11471-1-AP-150UL
 View more versions of this product
Product Code. 16848573
  • £349.00 / 150µL
Description
This gene encodes an EF-hand-containing calcium binding protein. The encoded protein likely plays a role in calcium homeostasis. Mutations in this gene have been associated with susceptibility to juvenile myoclonic epilepsy and juvenile absence epilepsy. Alternatively spliced transcript variants have been described.
Specifications
Specifications
EFHC1
Polyclonal
Unconjugated
EFHC1
EFHC1, EJM, EJM1, JAE, Myoclonin 1
Rabbit
Antigen Affinity Chromatography
RUO
114327, 301295, 71877
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5JVL4, Q9D9T8
Efhc1
EFHC1 Fusion Protein Ag2048
150 μL
Primary
Human, Mouse, Rat