missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Efficient-Pro™ Medium

Product Code. 30284822
Change view
Click to view available options
Quantity:
1000 mL
10 L
20 L
Unit Size:
1 L
10 L
20 L
Product Code. Quantity unitSize
30284822 10 L 10 L
30282666 1000 mL 1 L
30283683 20 L 20 L
3 options
This item is not returnable. View return policy

Product Code. 30284822

Brand: Gibco™ A5322202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco™ Efficient-Pro™ Media delivers superior product performance, improved product titer, better protein quality and lot-to-lot consistency for more control and predictability. Chemically defined and completely scalable, available in liquid or AGT™ formats for sustained cell viability.

  • A next generation CHO media used in the production of monoclonal antibodies
  • Liquid medium offers significant performance and ease-of-use improvements not seen in existing Gibco™ or competitor products
  • Formulated as a chemically-defined, animal-origin free cell culture media used in the bioproduction of mAbs & other therapeutics in traditional batch & fed-batch bioprocessing workflows
  • Free of growth factors and insulin to ensure batch-to-batch consistency and production efficiency
  • Designed to work optimally with Efficient-Pro Feed 1 and Efficient-Pro Feed 2 for use in CHO-K1 and DG-44/CHO-S cell lines respectively
  • The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles
  • System is easier to use with less solubility issues than other CHO media and feed offerings
  • Sustain cell viability for longer bioreactor runs & maximized titer
  • Demonstrated to improve titers in a CHO-K1 cell line up to 147% when paired with Efficient-Pro™ Feed 1 compared to an industry-leader’s media and feed system
  • Improve downstream outputs such as protein glycosylation, charge-variants & aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Increase flexibility with a range of format & packaging configurations
  • Decrease cell shock with simplified adaptation protocols
TRUSTED_SUSTAINABILITY

Specifications

Cell Line CHO-DG44, CHO-S, CHO-K1, GS-CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Quantity 10 L
Shipping Condition Room temperature
Green Features Sustainable packaging
Content And Storage Store in the dark at 2–8°C.

Culture Type Mammalian Cell Culture

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.