missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Efficient-Pro™ Feed Enhancer

Product Code. 30829974 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829974 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30829974 Supplier Gibco™ Supplier No. A4000135801
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Efficient-Pro Feed Enhancer is a liquid next-generation Chinese Hamster Ovary (CHO) feed enhancer used in the production of monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.

Designed to work optimally with Efficient-Pro Medium and Efficient-Pro Feed 3, Efficient-Pro Feed Enhancer has been formulated specifically for use in CHO-K1-GS cell lines. The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles, and is easier to use and scale with convenient formats available (liquid and DPM).

  • Sustain cell viability for longer bioreactor runs and maximized titer
  • Improve downstream outputs such as protein glycosylation, charge-variants, and aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Ease of use with fast hydration time to save time on workflow

Efficient-Pro Feed Enhancer has been formulated as a chemically defined, animal-origin-free cell culture feed supplement used in the bioproduction of mAbs and other therapeutics in traditional batch and fed-batch bioprocessing workflows, particularly in combination with CHO-K1 cell line variants. Efficient-Pro Feed Enhancer is a balanced formulation designed to meet the metabolic needs of mAb production and does not contain growth factors or insulin to support batch-to-batch consistency and production efficiency. It has been shown to improve productivity in a CHO-K1-GS cell line when paired with Efficient-Pro Feed 3 and Efficient-Pro Medium.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in the dark at 2°C to 8°C.
Cell Line CHO-K1 GS, CHO-K1
Cell Type Mammalian Cell Culture
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type CHO Media Supplement
Quantity 100 mL
Shipping Condition Room temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.