Learn More
Gibco™ Efficient-Pro™ Feed 3
Description
Designed to work optimally with Efficient-Pro Medium and Efficient-Pro Feed Enhancer, Efficient-Pro Feed 3 has been formulated specifically for use with CHO-K1-GS cell lines. The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles, and is easier to use and scale with convenient formats available (liquid and AGT).
Features of Efficient-Pro Feed 3 include:
- Sustain cell viability for longer bioreactor runs and maximized titer
- Improve downstream outputs such as protein glycosylation, charge-variants, and aggregation
- Speed-up process development and streamline transfer to manufacturing scale
- Increase flexibility with a range of format and packaging configurations
Efficient-Pro Feed 3 has been formulated as a chemically defined, animal-origin-free cell culture feed used in the bioproduction of mAbs and other therapeutics in traditional batch and fed-batch bioprocessing workflows, particularly in combination with CHO-K1 cell line variants. Efficient-Pro Feed 3 is a balanced formulation designed to meet the metabolic needs of mAb production and does not contain growth factors or insulin to support batch-to-batch consistency and production efficiency. It has been shown to improve productivity in a CHO-K1-GS cell line when paired with Efficient-Pro Feed Enhancer and Efficient-Pro Medium.
Specifications
Specifications
| Content And Storage | Store in the dark at 2°C to 8°C. |
| Cell Line | CHO-K1 GS, CHO-K1 |
| Cell Type | Mammalian Cell Culture |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Form | Liquid |
| Product Type | CHO Media Supplement |
| Quantity | 1 L |
| Shipping Condition | Room temperature |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.