missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Efficient-Pro™ DPM Feed Enhancer

Product Code. 30826343
Change view
Click to view available options
Quantity:
1 L
10 L
Unit Size:
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30826343 10 L 10 L
30827051 1 L -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826343 Supplier Gibco™ Supplier No. A4000175502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Efficient-Pro DPM Feed Enhancer is a dry-format next-generation Chinese Hamster Ovary (CHO) feed enhancer used in the production of monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.

Designed to work optimally with Efficient-Pro Medium and Efficient-Pro Feed 3, Efficient-Pro DPM Feed Enhancer has been formulated specifically for use in CHO-K1-GS cell lines. The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles, and is easier to use and scale with convenient formats available (liquid and DPM).

  • Increase ease-of-use with DPM dry format with simplified dissolution with minimal storage
  • Sustain cell viability for longer bioreactor runs and maximized titer
  • Improve downstream outputs such as protein glycosylation, charge-variants, and aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Ease of use with fast hydration time to save time on workflow

Efficient-Pro DPM Feed Enhancer has been formulated as a chemically defined, animal-origin-free cell culture feed supplement used in the bioproduction of mAbs and other therapeutics in traditional batch and fed-batch bioprocessing workflows, particularly in combination with CHO-K1 cell line variants. Efficient-Pro DPM Feed Enhancer is a balanced formulation designed to meet the metabolic needs of mAb production and does not contain growth factors or insulin to support batch-to-batch consistency and production efficiency. It has been shown to improve productivity in a CHO-K1-GS cell line when paired with Efficient-Pro Feed 3 and Efficient-Pro Medium.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in the dark at 2°C to 8°C.
Cell Line CHO-K1 GS, CHO-K1
Cell Type Mammalian Cell Culture
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Dehydrated, powder
Product Type CHO Media Supplement
Quantity 10 L
Shipping Condition Room temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.