missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EFEMP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106661
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EFEMP2.
EFEMP2 have been found to contain variations of the epidermal growth factor (EGF) domain and have been implicated in functions as diverse as blood coagulation, activation of complement and determination of cell fate during development. The protein contains four EGF2 domains and six calcium-binding EGF2 domains. This protein is necessary for elastic fiber formation and connective tissue development.
Specifications
| EFEMP2 | |
| Polyclonal | |
| Unconjugated | |
| Efemp2 | |
| 0610011K11Rik; ARCL1B; Efemp2; EGF containing fibulin extracellular matrix protein 2; EGF containing fibulin like extracellular matrix protein 2; EGF containing fibulin-like extracellular matrix protein 2; EGF-containing fibulin-like extracellular matrix protein 2; EGF-containing fibulin-like extracellular matrix protein 2 (Efemp2); epidermal growth factor-containing fibulin-like extracellular matrix protein 2; Fbln4; FIBL-4; fibulin 4; Fibulin4; fibulin-4; Mbp1; mutant p53 binding protein 1; mutant p53-binding protein 1; Protein UPH1; UNQ200/PRO226; UPH1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 30008 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95967 | |
| Efemp2 | |
| A synthesized peptide derived from human EFEMP2(Accession O95967), corresponding to amino acid residues G93-L143. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction