missing translation for 'onlineSavingsMsg'
Learn More

EFEMP2 Antibody, Novus Biologicals™

Product Code. 18372229 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372229 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372229 Supplier Novus Biologicals Supplier No. H00030008B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

EFEMP2 Polyclonal antibody specifically detects EFEMP2 in Human samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFEMP2
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry
Formulation PBS (pH 7.4)
Gene Accession No. AAH10456.1
Gene Alias EGF containing fibulin-like extracellular matrix protein 2, EGF-containing fibulin-like extracellular matrix protein 2, FBLN4fibulin-4, FIBL-4, fibulin 4, Fibulin-4, MBP1, Protein UPH1, UPH1mutant p53 binding protein 1
Host Species Mouse
Immunogen EFEMP2 (AAH10456.1, 1 a.a. - 443 a.a.) full-length human protein. MLPCASCLPGSLLLWALLLLLLGSASPQDSEEPDSYTECTDGYEWDPDSQHCRDVNECLTIPEACKGEMKCINHYGGYLCLPRSAAVINDLHGEGPPPPVPPAQHPNPCPPGYEPDDQDSCVDVDECAQALHDCRPSQDCHNLPGSYQCTCPDGYRKIGPECVDIDECRYRYCQHRCVNLPGSFRCQCEPGFQLGPNNRSCVDVNECDMGAPCEQRCFNSYGTFLCRCHQGYELHRDGFSCSDIDECSYSSYLCQYRCVNEPGRFSCHCPQGYQLLATRLCQDIDECESGAHQCSEAQTCVNFHGGYRCVDTNRCVEPYIQVSENRCLCPASNPLCREQPSSIVHRYMTITSERSVPADVFQIQATSVYPGAYNAFQIRAGNSQGDFYIRQINNVSAMLVLARPVTGPREYVLDLEMVTMNSLMSYRASSVLRLTVFVGAYTF
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30008
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.