missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFCAB6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546780
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: FGLKATTKIN WKQFLTSFHE PQGLQVSSKG PLTKRNSINS RNESHKENII
EFCAB6 Polyclonal antibody specifically detects EFCAB6 in Human samples. It is validated for Western Blot.Specifications
EFCAB6 | |
Polyclonal | |
Unconjugated | |
EFCAB6 | |
CAP-binding protein complex-interacting protein 1; dJ185D5.1; DJ-1-binding protein; DJBP; EFCAB6; EF-hand calcium binding domain 6; EF-hand calcium-binding domain-containing protein 6; HSCBCIP1; KIAA1672 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64800 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5THR3 | |
EFCAB6 | |
Synthetic peptide directed towards the N-terminal of human EFCAB6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |