missing translation for 'onlineSavingsMsg'
Learn More

EFCAB6 Antibody, Novus Biologicals™

Product Code. 18417600 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417600 0.1 mL 0.1mL
18467510 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18417600 Supplier Novus Biologicals Supplier No. NBP184237

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EFCAB6 Polyclonal antibody specifically detects EFCAB6 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias CAP-binding protein complex interacting protein 1, dJ185D5.1, DJBPCAP-binding protein complex-interacting protein 1, EF-hand calcium binding domain 6, EF-hand calcium-binding domain-containing protein 6, FLJ23588, HSCBCIP1, KIAA1672DJ-1-binding protein
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:QQSTEKAVAARDKLMDRHQDISKAFTKTDQSKTNYISICKMQEVLEECGCSLTEGELTHLLNSWGVSRHDNAINYLDFLRAVENSKSTGAQPKEKEESMPINFATLNPQEVVRKIQEVVESSQLALSTAFSALDKEDT
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64800
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.