Learn More
EFCAB4A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561187
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LEAVFESLDQ AHTGFLTARE FCLGLGMFVG VASAQGANPC RTPEETFESG GLDVQGTAGS LDEE Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.
EFCAB4A, also known as Calcium release-activated calcium channel regulator 2B, is a novel Ca2+-binding EF-hand protein that is thought to play a key role in store-operated Ca2+ entry in T-cells by regulating CRAC channel activation, but the detailed function is still under investigation. It is likely to play a similar role as the related protein EFCAB4B, which acts as a cytoplasmic calcium-sensor that forms a complex with ORAI1 and STIM1 at the junctional regions between the plasma membrane and the endoplasmic reticulum upon low Ca2+ concentration.Specifications
EFCAB4A | |
Polyclonal | |
Unconjugated | |
CRACR2B | |
6330520A15Rik; BC026645; Ca2+ release-activated Ca2+ (CRAC) channel regulator 2B; calcium release activated channel regulator 2B; calcium release-activated calcium channel regulator 2B; Calcium release-activated channel regulator 2B; CRAC channel regulator 2B; CRAC regulator 2B; Cracr2a; CRACR2B; Efcab4; EFCAB4A; EF-hand calcium binding domain 4A; EF-hand calcium-binding domain-containing protein 4A; RGD1560911 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N4Y2 | |
CRACR2B | |
Recombinant protein corresponding to Human EFCAB4A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only