missing translation for 'onlineSavingsMsg'
Learn More

EFCAB4A Polyclonal Antibody

Product Code. 12859202
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12859202 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12859202 Supplier Invitrogen Supplier No. PA521059

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EFCAB4A Polyclonal Antibody

EFCAB4A Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB4A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CRACR2B
Gene Accession No. B0BNK9, Q80ZJ8, Q8N4Y2
Gene Alias 6330520A15Rik; BC026645; Ca2+ release-activated Ca2+ (CRAC) channel regulator 2B; calcium release activated channel regulator 2B; calcium release-activated calcium channel regulator 2B; Calcium release-activated channel regulator 2B; CRAC channel regulator 2B; CRAC regulator 2B; Cracr2a; CRACR2B; Efcab4; EFCAB4A; EF-hand calcium binding domain 4A; EF-hand calcium-binding domain-containing protein 4A; RGD1560911
Gene Symbols CRACR2B
Host Species Rabbit
Immunogen A 15 amino acid peptide near the center of human EFCAB4A.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213573, 283229, 309112
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.