missing translation for 'onlineSavingsMsg'
Learn More

EFCAB4A Antibody, DyLight 350, Novus Biologicals™

Product Code. 18400297 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400297 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400297 Supplier Novus Biologicals Supplier No. NBP176492UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EFCAB4A Polyclonal antibody specifically detects EFCAB4A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB4A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Calcium release-activated calcium channel regulator 2B, CRAC channel regulator 2B, CRACR2B, EF-hand calcium binding domain 4A, EF-hand calcium-binding domain-containing protein 4A, FLJ46033, MGC45840, MGC88858
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the center of human EFCAB4A. The immunogen is located within amino acids 290 - 340 of EFCAB4A. Amino Acid Squence: ESEARGRQEQTQRDV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 283229
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.