missing translation for 'onlineSavingsMsg'
Learn More

Efavirenz 1-Desamine-1-oxy, TRC

Product Code. 30380685
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
100mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380685 10 mg 10mg
30375812 1 mg 1mg
30373813 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380685 Supplier Toronto Research Chemicals Supplier No. TRCE42503510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Efavirenz 1-Desamine-1-oxy
Analyte or Component Names Efavirenz 1-Desamine-1-oxy
Shipping Condition Room Temperature
Molecular Formula C14H8ClF3O3
Synonym 6-Chloro-4-(cyclopropylethynyl)-4-(trifluoromethyl)-4H-benzo[d][1,3]dioxin-2-one
InChI Formula InChI=1S/C13H16O5/c1-3-17-11-7-9(8-12(14)15)5-6-10(11)13(16)18-4-2/h5-7H,3-4,8H2,1-2H3,(H,14,15)
Solution Type Neat
SMILES CCOC(=O)c1c(OCC)cc(CC(O)=O)cc1
Industry Type Pharmaceutical
Molecular Weight (g/mol) 316.66
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.