missing translation for 'onlineSavingsMsg'
Learn More

Efaproxiral sodium, MedChemExpress

Product Code. 16401937
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401937 Supplier Medchem Express Supplier No. HY13619A
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Efaproxiral sodium (RSR13 sodium) is a synthetic allosteric modifier of haemoglobin (Hb), decreases Hb-oxygen (O2) binding affinity and enhances oxygenation of hypoxic tumours during radiation therapy.

Efaproxiral sodium (RSR13 sodium) is a synthetic allosteric modifier of haemoglobin (Hb), decreases Hb-oxygen (O2) binding affinity and enhances oxygenation of hypoxic tumours during radiation therapy.

Specifications

Chemical Name or Material Efaproxiral sodium
CAS 170787-99-2
Color White
Purity Grade Notes Research
Molecular Formula C20H22NNaO4
Quantity 50 mg
Synonym RSR13 sodium
Solubility Information DMSO : ≥ 39 mg/mL (107.33 mM)
SMILES CC(C)(OC1=CC=C(CC(NC2=CC(C)=CC(C)=C2)=O)C=C1)C(O[Na])=O
Molecular Weight (g/mol) 363.38
Formula Weight 363.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.