missing translation for 'onlineSavingsMsg'
Learn More

Efaproxiral, MedChemExpress

Product Code. 16491927
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16491927

Brand: Medchem Express HY13619

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Efaproxiral is a haemoglobin (Hb) synthetic allosteric modifier, decreases Hb-oxygen (O2) binding affinity and enhances oxygenation of hypoxic tumours during radiation therapy .

Efaproxiral is a haemoglobin (Hb) synthetic allosteric modifier, decreases Hb-oxygen (O2) binding affinity and enhances oxygenation of hypoxic tumours during radiation therapy .

Specifications

Chemical Name or Material Efaproxiral
CAS 131179-95-8
Color White
Purity Grade Notes Research
Molecular Formula C20H23NO4
Quantity 50 mg
Synonym RSR13
Solubility Information DMSO : ≥ 150 mg/mL (439.37 mM)
SMILES CC(C)(OC1=CC=C(CC(NC2=CC(C)=CC(C)=C2)=O)C=C1)C(O)=O
Molecular Weight (g/mol) 341.4
Formula Weight 341.4
Percent Purity 98.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.