missing translation for 'onlineSavingsMsg'
Learn More

EF-5, MedChemExpress

Product Code. 16432378
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16432378 5 mg 5mg
16473159 1 mg 1mg
16430179 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16432378 Supplier Medchem Express Supplier No. HYU00118
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress EF-5 (EF5; 2-Nitroimidazole) is a hypoxia labeling agent used to identify hypoxia in cells.

EF-5 (EF5

Specifications

Chemical Name or Material EF-5
CAS 152721-37-4
Color White
Purity Grade Notes Research
Molecular Formula C8H7F5N4O3
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (413.69 mM; Need ultrasonic)
SMILES O=C(NCC(F)(F)C(F)(F)F)CN1C=CN=C1[N+]([O-])=O
Molecular Weight (g/mol) 302.16
Formula Weight 302.16
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.