missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Eeyarestatin I
GREENER_CHOICE

Product Code. 18014892
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18014892

Brand: Tocris Bioscience™ 3922/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent inhibitor of ER-associated protein degradation and translocation

Eeyarestatin I is a potent inhibitor of endoplasmic reticulum associated protein degradation (ERAD). Specifically targets the p97-associated deubiquitinating process (PAD) and inhibits ataxin-3 (atx3)-dependent deubiquitination. Also inhibits Sec61-mediated protein translocation at the ER. Displays cytotoxic activity preferentially against cancer cells; induces cell death via the proapoptotic protein NOXA. Also enhances adeno-associated viral transduction in multiple cells types.
TRUSTED_SUSTAINABILITY

Specifications

CAS 412960-54-4
Quantity 10 mg
Molecular Formula C27H25Cl2N7O7
Formula Weight 630.44
Purity >98%
Chemical Name or Material Eeyarestatin I
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.