missing translation for 'onlineSavingsMsg'
Learn More

EEPD1 Antibody, Novus Biologicals™

Product Code. 18423109 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423109 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423109 Supplier Novus Biologicals Supplier No. H00080820B03P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

EEPD1 Polyclonal antibody specifically detects EEPD1 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEPD1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH65518.1
Gene Alias endonuclease/exonuclease/phosphatase family domain containing 1, endonuclease/exonuclease/phosphatase family domain-containing protein 1, KIAA1706HSPC107
Host Species Mouse
Immunogen KIAA1706 (AAH65518.1, 1 a.a. - 569 a.a.) full-length human protein. MGSTLGCHRSIPRDPSDLSHSRKFSAACNFSNILVNQERLNINTATEEELMTLPGVTRAVARSIVEYREYIGGFKKVEDLALVSGVGATKLEQVKFEICVSSKGSSAQHSPSSLRRDLLAEQQPHHLATAVPLTPRVNINTATPAQLMSVRGLSEKMALSIVDFRREHGPFRSVEDLVRMDGINAAFLDRIRHQVFAERSRPPSTHTNGGLTFTAKPHPSPTSLSLQSEDLDLPPGGPTQIISTRPSVEAFGGTRDGRPVLRLATWNLQGCSVEKANNPGVREVVCMTLLENSIKLLAVQELLDREALEKFCTELNQPTLPNIRKWKGPRGCWKAVVAEKPSNQLQKGAGYAGFLWDAAAGMELRDAGSQESSPSNGHGKLAGPSPYLGRFKVGSHDLTLVNLHLAALTLLGSENPSKNHSDGHRLASFAQTLQETLKGEKDVIILGDFGQGPDSNDYDILRKEKFHHLIPAHTFTNISTKNPQGSKSLDNIWISKSLKKVFTGHWAVVREGLTNPWIPDNWSWGGVASEHCPVLAEFYTEKDWSKKDAPRNGSGVALERSEANIKHER
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80820
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.