missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38429-100ul
This item is not returnable.
View return policy
Description
EEN Polyclonal antibody specifically detects EEN in Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| EEN | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CNSA1endophilin-2, EEN fusion partner of MLL, EENSH3 domain-containing GRB2-like protein 1, Endophilin-2, extra 11-19 leukemia fusion, Extra eleven-nineteen leukemia fusion gene protein, MGC111371, SH3 domain protein 2B, SH3-containing Grb-2-like 1 protein, SH3D2Bendophilin-A2, SH3-domain GRB2-like 1, SH3P8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 219-368 of human EEN (NP_003016.1).,, Sequence:, VDAQLDYHRQAVQILDELAEKLKRRMREASSRPKREYKPKPREPFDLGEPEQSNGGFPCTTAPKIAASSSFRSSDKPIRTPSRSMPPLDQPSCKALYDFEPENDGELGFHEGDVITLTNQIDENWYEGMLDGQSGFFPLSYVEVLVPLPQ | |
| 100 μL | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6455 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction