Learn More
Invitrogen™ EEF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527662
Description
Recommended positive controls: 293T, A431, Neuro2A, NIH-3T3, Neuro 2A, Raw264.7, PC-12. Predicted reactivity: Mouse (98%), Rat (98%), Chicken (95%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catalyzes the GTP-dependent ribosomal translocation step during translation elongation. During this step, the ribosome changes from the pre-translocational (PRE) to the post-translocational (POST) state as the newly formed A-site-bound peptidyl-tRNA and P-site-bound deacylated tRNA move to the P and E sites, respectively. Catalyzes the coordinated movement of the two tRNA molecules, the mRNA and conformational changes in the ribosome.
Specifications
| EEF2 | |
| Polyclonal | |
| Unconjugated | |
| EEF2 | |
| EEF2; EEF-2; Ef2; EF-2; Elongation factor 2; elongation factor-2; eukaryotic translation elongation factor 2; I79_013848; polypeptidyl-tRNA translocase; SCA26; zef2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13629, 1938, 29565 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.86 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P05197, P13639, P58252 | |
| EEF2 | |
| Recombinant protein encompassing a sequence within the center region of human eEF2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.