Learn More
EEF2 Mouse anti-Human, Unconjugated, Clone: 1E4D5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67550-1-IG-20UL
Description
The EEF2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EEF2. This antibody recognizes human, mouse, rat antigen. The EEF2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| EEF2 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1E4D5 | |
| Unconjugated | |
| 20 μL | |
| 1938 | |
| IgG1 |