Learn More
EEF2 Mouse, anti-Human, CoraLite 488, Clone: 1E4D5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67550-100UL
Description
The EEF2 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human EEF2. This antibody recognizes human, mouse, rat antigen. The EEF2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| EEF2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |
| Immunofluorescence | |
| 1E4D5 | |
| CoraLite 488 | |
| 100 μL | |
| 1938 | |
| Liquid |