missing translation for 'onlineSavingsMsg'
Learn More

EEF1G Rabbit anti-Human, Mouse, Rat, Clone: 2E1O7, Novus Biologicals™

Product Code. 18341515 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341515 100 μg 100µL
18338441 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18341515 Supplier Novus Biologicals Supplier No. NBP316694100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EEF1G Monoclonal antibody specifically detects EEF1G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen EEF1G
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2E1O7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 to 4 μg/mL, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias eEF-1B gamma, EF-1-gamma, EF1Gelongation factor 1-gamma, eukaryotic translation elongation factor 1 gamma, GIG35, pancreatic tumor-related protein, PRO1608, translation elongation factor eEF-1 gamma chain
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-123 of human EEF1G (P26641).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1937
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.