missing translation for 'onlineSavingsMsg'
Learn More

EEF1G Antibody (S1), Novus Biologicals™

Product Code. 18353858 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353858 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353858 Supplier Novus Biologicals Supplier No. H00001937M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EEF1G Monoclonal antibody specifically detects EEF1G in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEF1G
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone S1
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH15813
Gene Alias eEF-1B gamma, EF-1-gamma, EF1Gelongation factor 1-gamma, eukaryotic translation elongation factor 1 gamma, GIG35, pancreatic tumor-related protein, PRO1608, translation elongation factor eEF-1 gamma chain
Host Species Mouse
Immunogen EEF1G (AAH15813.1, 1 a.a. ~ 437 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAGTLYTYPENWRAFKALIAAQYSGAQVRVLSAPPHFHFGQTNRTPEFLRKFPAGKVPAFEGDDGFCVFESNAIAYHVSNEELRGSTPEAAAQVVQWVSFADSDIVPPASTWVFPTLGIMHHDKQATENAKEEVRRILGLLDAYLKTRTFLVGERVTLADITVVCTLLWLYKQVLEPSFRQAFPNTNRWFLTCINQPQFRAVLGEVKLCEKMAQFDAKKFAETQPKKDTPRKEKGSREEKQKPQAERKEEKKAAAPAPEEEMDECEQALAAEPKAKDPFAHLPKSTFVLDEFKRKYSNEDTLSVALPYFWEHFDKDGWSLWYSEYRFPEELTQTFMSCNLITGMFQRLDKLRKNAFASVILFGTNNSSSISGVWVFRGQELAFPLSPDWQVDYESYTWRKLDPGSEETQTLVREYFSWEGAFQHVGKAFNQGKIFK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1937
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.