missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1G Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38481-100ul
This item is not returnable.
View return policy
Description
EEF1G Polyclonal antibody specifically detects EEF1G in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| EEF1G | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| eEF-1B gamma, EF-1-gamma, EF1Gelongation factor 1-gamma, eukaryotic translation elongation factor 1 gamma, GIG35, pancreatic tumor-related protein, PRO1608, translation elongation factor eEF-1 gamma chain | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 168-437 of human EEF1G (NP_001395.1).,, Sequence:, LLWLYKQVLEPSFRQAFPNTNRWFLTCINQPQFRAVLGEVKLCEKMAQFDAKKFAETQPKKDTPRKEKGSREEKQKPQAERKEEKKAAAPAPEEEMDECEQALAAEPKAKDPFAHLPKSTFVLDEFKRKYSNEDTLSVALPYFWEHFDKDGWSLWYSEYRFPEELTQTFMSCNLITGMFQRLDKLRKNAFASVILFGTNNSSSISGVWVFRGQELAFPLSPDWQVDYESYTWRKLDPGSEETQTLVREYFSWEGAFQHVGKAFNQGKIFK | |
| 100 μL | |
| Cell Cycle and Replication | |
| 1937 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction