missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10805-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a multifunctional protein that localizes both in the cytoplasm and in the nucleus. In the cytoplasm, the encoded protein is an auxiliary component of the macromolecular aminoacyl-tRNA synthase complex. However, its mouse homolog has been shown to translocate to the nucleus in response to DNA damage and plays a positive role in ATM/ATR-mediated p53 activation. Alternative splicing results in multiple transcript variants.Specifications
| EEF1E1 | |
| Polyclonal | |
| Unconjugated | |
| EEF1E1 | |
| AIMP3, EEF1E1, Elongation factor p18, P18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291057, 66143, 9521 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43324, Q9D1M4 | |
| Eef1e1 | |
| EEF1E1 Fusion Protein Ag1092 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction