missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11402-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human EEF1A1.
This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis. With PARP1 and TXK, forms a complex that acts as a T helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.Specifications
| EEF1A1 | |
| Polyclonal | |
| Unconjugated | |
| EEF1A1 | |
| CCS 3, CCS3, EEF 1, EEF1A, eEF1A 1, EEF1A1, EF 1 alpha 1, EF Tu, EF1A, Elongation factor 1 alpha 1, Elongation factor Tu, GRAF 1EF, HNGC:16303, LENG7, PTI1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13627, 171361, 1915 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P10126, P62630, P68104 | |
| Eef1a1 | |
| EEF1A1 Fusion Protein Ag1938 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction