missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EEF1A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584080
This item is not returnable.
View return policy
Description
Immunogen sequence: LLEALDTILP PTRPTDKPLR LPLQDVYKIG GIGTVPVGRV ETGIL Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis. With PARP1 and TXK, forms a complex that acts as a T helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.
Specifications
| EEF1A1 | |
| Polyclonal | |
| Unconjugated | |
| Eef1a1 | |
| CCS3; CCS-3; cervical cancer suppressor 3; CTCL tumor antigen; EE1A1; EEF-1; EEF1A; eef1a1; eEF1A-1; eef1a1.S; Eef1a2; Eef1a2l1; eef1as; eef1a-s; EF-1 alpha; EF-1 alpha2; EF1A; EF1A1; EF1a-like protein; EF1alpha; EF-1-alpha-1; EF-1-alpha-S; EF-Tu; elongation factor 1 A-1; elongation factor 1 alpha subunit; elongation factor 1-alpha 1; Elongation factor 1-alpha, somatic form; elongation factor Tu; elongation factor-1 alpha; eukaryotic elongation factor 1 A-1; eukaryotic translation elongation factor 1 alpha 1; eukaryotic translation elongation factor 1 alpha 1 S homeolog; eukaryotic translation elongation factor 1 alpha 1-like 14; eukaryotic translation elongation factor 1 alpha 2 like 1; eukaryotic translation elongation factor 1 alpha, somatic form; glucocorticoid receptor AF-1 specific elongation factor; GRAF 1EF; GRAF-1EF; HNGC:16303; LENG7; leukocyte receptor cluster (LRC) member 7; leukocyte receptor cluster member 7; prostate tumor-inducing protein 1; PTI1; SI; translation elongation factor 1 alpha 1-like 14; XELAEV_18029462mg | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1915 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P68104 | |
| Eef1a1 | |
| Recombinant protein corresponding to Human EEF1A1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction