missing translation for 'onlineSavingsMsg'
Learn More

EED226, MedChemExpress

Product Code. 16469816
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469816 50 mg 50mg
16455997 5 mg 5mg
16433278 10 mg 10mg
16424536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16469816 Supplier Medchem Express Supplier No. HY101117
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress EED226 is a polycomb repressive complex 2 (PRC2) inhibitor, which binds to the K27me3-pocket on embryonic ectoderm development (EED) and shows strong antitumor activity in xenograft mice model. EED226 is a potent, selective, and orally bioavailable EED inhibitor. EED226 inhibits PRC2 with an IC50 of 23.4 nM when the H3K27me0 peptide is used as a substrate in the in vitro enzymatic assays.

EED226 is a polycomb repressive complex 2 (PRC2) inhibitor, which binds to the K27me3-pocket on embryonic ectoderm development (EED) and shows strong antitumor activity in xenograft mice model. EED226 is a potent, selective, and orally bioavailable EED inhibitor. EED226 inhibits PRC2 with an IC50 of 23.4 nM when the H3K27me0 peptide is used as a substrate in the in vitro enzymatic assays.

Specifications

Chemical Name or Material EED226
CAS 2083627-02-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H15N5O3S
Quantity 50 mg
Solubility Information DMSO : ≥ 29 mg/mL (78.51 mM)
SMILES O=S(C1=CC=C(C2=CN=C(NCC3=CC=CO3)N4C2=NN=C4)C=C1)(C)=O
Molecular Weight (g/mol) 369.4
Formula Weight 369.4
Percent Purity 98.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.