missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EED Monoclonal Antibody (K06320_10A3), Invitrogen™

Mouse Monoclonal Antibody

£416.00 - £734.00

Specifications

Antigen EED
Clone K06320_10A3
Concentration 1 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30204481
View Documents
Invitrogen™
MA556061
200 μL
£734.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30199801
View Documents
Invitrogen™
MA555681
50 μL
£416.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: AGTDMPAAKK QKLSSDENSN PDLSGDENDD AVSIESGTNT ERPDTPTNTP NAPGRKSWGK GKWKSKKCKY SFKCVNSLKE DHNQPLFGVQ FNWHSKEGDP LVFATVGSNR VTLYECHSQG EIRLLQSYVD ADADENFYTC

EED Monoclonal antibody specifically detects EED in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot
Specifications

Specifications

EED
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
EED; embryonic ectoderm development; Embryonic ectoderm development protein; ENSMUSG00000039373; HEED; l(7)5Rn; l7Rn5; lethal, Chr 7, Rinchik 5; lusk; Polycomb protein EED; WAIT1; WAIT-1; WD protein associating with integrin cytoplasmic tails 1
Recombinant human EED (immunogen range is 11-150).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
EED
K06320_10A3
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human, Mouse
O75530, Q921E6
13626, 8726
IgG1
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.