missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EED Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98765PE
This item is not returnable.
View return policy
Description
EED Polyclonal antibody specifically detects EED in Human samples. It is validated for Western Blot
Specifications
| Western Blot | |
| Antigen and protein A Affinity-purified | |
| DNA replication Transcription Translation and Splicing | |
| Store at 4C in the dark. |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human EED | |
| 0.1 mL | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction