missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EED Polyclonal antibody specifically detects EED in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | EED |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence |
| Host Species | Rabbit |
| Immunogen | EED antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human EED. The immunogen is located within amino acids 30 - 80 of EED. Amino Acid Squence: ESGTNTERPDTPTNTPNA |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?