missing translation for 'onlineSavingsMsg'
Learn More

EED Antibody, Allophycocyanin, R&D Systems™

Product Code. 18604915 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18604915 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18604915 Supplier Novus Biologicals Supplier No. NBP298765APC
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

EED Polyclonal specifically detects EED in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human EED
Product Type Antibody
Isotype IgG
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.