missing translation for 'onlineSavingsMsg'
Learn More

EEA1 Antibody (2G2), Novus Biologicals™

Product Code. 18389118 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389118 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389118 Supplier Novus Biologicals Supplier No. H00008411M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EEA1 Monoclonal antibody specifically detects EEA1 in Human, Mouse, Hamster samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEA1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2G2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003557
Gene Alias early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2
Host Species Mouse
Immunogen EEA1 (NP_003557, 1312 a.a. ~ 1411 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KVLELQRKLDNTTAAVQELGRENQSLQIKHTQALNRKWAEDNEVQNCMACGKGFSVTVRRHHCRQCGNIFCAECSAKNALTPSSKKPVRVCDACFNDLQG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Endosome Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 8411
Target Species Human, Mouse, Hamster
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.