missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Edwards Medium with Sheep Blood

Product Code. 10017212
Change view
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10017212 10 Plates Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10017212 Supplier Thermo Scientific™ Supplier No. PB5080A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Unlock the power of precision in microbial cultivation with PB5080A Edwards Medium with Sheep Blood.

This high-performance agar medium is specifically designed to support the growth and isolation of a wide range of microorganisms, ensuring accurate and reliable results for your microbiological testing needs.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage 2°C to 12°C
Format 90 mm
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Volume 17 mL
Packaging Type 1 x 10 Plates
Quantity 10 Plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.