missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid tetrasodium salt dihydrate, (EDTA), 99-101%, Ultrapure

Product Code. 15895278 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
25 kg
Packaging:
Plastic bottle
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15895278 1 kg Plastic bottle 1kg
15805288 25 kg Plastic bottle 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15895278 Supplier Thermo Scientific Supplier No. J15700.A1
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10378-23-1 ∣ C10H20N2Na4O10 ∣ 420.233 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-23-1
Molecular Formula C10H20N2Na4O10
Molecular Weight (g/mol) 420.233
MDL Number MFCD00150026
InChI Key DGWOWDJDIBPYOR-UHFFFAOYSA-N
Synonym ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate
PubChem CID 131843362
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na]

Specifications

Physical Form Crystalline Powder
Quantity 1 kg
Beilstein 3861753
Formula Weight 416.2
Percent Purity 99 to 101%
Grade Ungraded
Packaging Plastic bottle
Chemical Name or Material Ethylenediaminetetraacetic Acid Tetrasodium Salt Dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.