missing translation for 'onlineSavingsMsg'
Learn More

EDTA, Sodium Ferric Salt, MP Biomedicals

Product Code. 11402735 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402735 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11402735 Supplier MP Biomedicals Supplier No. 0520890350
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chelating agent used to eliminate enzyme inhibition by traces of heavy metals and to inhibit enzymes requiring divalent cations as cofactors.

Specifications

CAS 15708-41-5
Molecular Formula C10H12FeN2NaO8
Quantity 50 g
Synonym sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edta
InChI Key MKWYFZFMAMBPQK-UHFFFAOYSA-J
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3]
IUPAC Name sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+)
Molecular Weight (g/mol) 367.047
PubChem CID 27461
ChEBI CHEBI:78292
Formula Weight 367.05
Grade Not Available
Physical Form Powder
Chemical Name or Material EDTA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.