missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium zinc salt tetrahydrate, Honeywell Fluka™

Product Code. 16643954 Shop All Honeywell Fluka Products
Click to view available options
:
25 kg
This item is not returnable. View return policy

Product Code. 16643954

Brand: Honeywell Fluka 3455325KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 176736-49-5
CAS Max % 1
Molecular Formula C10H20N2Na2O12Zn
Linear Formula C10H12N2Na2O8Zn · 4H2O
Quantity 25 kg
Synonym Disodium zinc ethylenediaminetetraacetate tetrahydrate; EDTA-Na2Zn
InChI Key ZEVLVCFKKCNROJ-UHFFFAOYSA-J
SMILES O.O.O.O.[Na+].[Na+].[Zn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
IUPAC Name zinc(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate
Molecular Weight (g/mol) 471.63
Grade Reagent Grade Salts
Chemical Name or Material EDTA disodium zinc salt tetrahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.